Assay Details
Gene Symbol: pnr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275691.2 | NP_001262620.1 | 3 - 4 | 1409 | 61 | |
NM_001275692.2 | NP_001262621.1 | 3 - 4 | 1657 | 61 | ||
NM_057337.3 | NP_476685.1 | 3 - 4 | 1418 | 61 | ||
NM_169697.3 | NP_732102.1 | 3 - 4 | 1484 | 61 | ||
GenBank mRNA | BT031157.1 | - | 3 - 4 | 1416 | 61 | |
BT053737.1 | - | 3 - 4 | 1481 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
