Assay Details
Gene Symbol: ss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170158.3 | NP_001163629.2 | 2 - 3 | 763 | 79 | |
NM_001260211.2 | NP_001247140.1 | 2 - 3 | 785 | 79 | ||
NM_057400.4 | NP_476748.1 | 2 - 3 | 800 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
