Assay Details
Gene Symbol: ss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170158.3 | NP_001163629.2 | 3 - 4 | 944 | 91 | |
NM_001260211.2 | NP_001247140.1 | 3 - 4 | 966 | 91 | ||
NM_057400.4 | NP_476748.1 | 3 - 4 | 981 | 91 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
