Assay Details
| Gene Symbol: awd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300695.1 | NP_001287624.1 | 1 - 2 | 163 | 65 | |
| NM_001300696.1 | NP_001287625.1 | 1 - 2 | 312 | 65 | ||
| NM_057413.4 | NP_476761.3 | 1 - 2 | 219 | 65 | ||
| GenBank mRNA | AY113576.1 | - | 1 - 2 | 219 | 65 | |
| KX531763.1 | - | 1 - 2 | 133 | 65 | ||
| X13107.1 | - | 158 | 65 | |||
More Information
Set Membership: |
|

awd
Genomic Map