Assay Details
| Gene Symbol: trx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014621.2 | NP_001014621.1 | 3 - 4 | 2076 | 65 | |
| NM_057421.3 | NP_476769.1 | 4 - 5 | 3313 | 65 | ||
| NM_057422.3 | NP_476770.1 | 4 - 5 | 2525 | 65 | ||
| NM_134281.2 | NP_599108.1 | 3 - 4 | 2076 | 65 | ||
| NM_134282.2 | NP_599109.1 | 5 - 6 | 3762 | 65 | ||
More Information
Set Membership: |
|

trx
Genomic Map