Assay Details
| Gene Symbol: trx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014621.2 | NP_001014621.1 | 6 - 7 | 10621 | 64 | |
| NM_057421.3 | NP_476769.1 | 7 - 8 | 11858 | 64 | ||
| NM_057422.3 | NP_476770.1 | 7 - 8 | 11070 | 64 | ||
| NM_134281.2 | NP_599108.1 | 6 - 7 | 10621 | 64 | ||
| NM_134282.2 | NP_599109.1 | 8 - 9 | 12307 | 64 | ||
More Information
Set Membership: |
|

trx
Genomic Map