Assay Details
| Gene Symbol: Hrb87F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170131.2 | NP_001163602.1 | 3 - 4 | 1301 | 75 | |
| NM_057458.5 | NP_476806.2 | 3 - 4 | 1301 | 75 | ||
| NM_057459.5 | NP_476807.1 | 3 - 4 | 1301 | 75 | ||
| GenBank mRNA | BT012315.1 | - | 3 - 4 | 1272 | 75 | |
More Information
Set Membership: |
|

Hrb87F
Genomic Map