Assay Details
Gene Symbol: CycC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057500.4 | NP_476848.1 | 1 - 2 | 736 | 59 | |
GenBank mRNA | AY061425.1 | - | 1 - 2 | 706 | 59 | |
X62948.1 | - | 652 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
