Assay Details
| Gene Symbol: wdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057552.4 | NP_476900.1 | 2 - 3 | 2474 | 62 | |
| GenBank mRNA | AY058690.1 | - | 2 - 3 | 2449 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wdn| Gene Symbol: wdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057552.4 | NP_476900.1 | 2 - 3 | 2474 | 62 | |
| GenBank mRNA | AY058690.1 | - | 2 - 3 | 2449 | 62 | |
Set Membership: |
|