Assay Details
Gene Symbol: Nmdmc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275469.1 | NP_001262398.1 | 2 - 3 | 959 | 63 | |
NM_057581.5 | NP_476929.1 | 2 - 3 | 983 | 63 | ||
NM_057582.5 | NP_476930.1 | 2 - 3 | 882 | 63 | ||
GenBank mRNA | AY047498.1 | - | 2 - 3 | 957 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
