Assay Details
| Gene Symbol: Dop1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170136.2 | NP_001163607.1 | 1 - 2 | 818 | 70 | |
| NM_001260163.1 | NP_001247092.1 | 1 - 2 | 818 | 70 | ||
| NM_001275634.1 | NP_001262563.1 | 1 - 2 | 818 | 70 | ||
| NM_001316525.1 | NP_001303454.1 | 1 - 2 | 818 | 70 | ||
| NM_057659.4 | NP_477007.2 | 1 - 2 | 818 | 70 | ||
More Information
Set Membership: |
|

Dop1R1
Genomic Map