Assay Details
| Gene Symbol: mRpL47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176433.2 | NP_788610.1 | 1 - 2 | 148 | 83 | |
| GenBank mRNA | AY089479.1 | - | 1 - 2 | 347 | 83 | |
More Information
Set Membership: |
|

mRpL47
Genomic Map