Assay Details
| Gene Symbol: noi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057766.5 | NP_477114.1 | 1 | 1782 | 140 | |
| GenBank mRNA | AY051676.1 | - | 1 | 1762 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

noi| Gene Symbol: noi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057766.5 | NP_477114.1 | 1 | 1782 | 140 | |
| GenBank mRNA | AY051676.1 | - | 1 | 1762 | 140 | |
Set Membership: |
|