Assay Details
Gene Symbol: Apc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276117.1 | NP_001263046.1 | 2 - 3 | 3996 | 64 | |
NM_001316494.1 | NP_001303423.1 | 2 - 3 | 4120 | 64 | ||
NM_057804.3 | NP_477152.1 | 2 - 3 | 4120 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
