Assay Details
Gene Symbol: Abi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104315.2 | NP_001097785.1 | 2 - 3 | 1126 | 65 | |
NM_001260162.1 | NP_001247091.1 | 3 - 4 | 1132 | 65 | ||
NM_001275631.1 | NP_001262560.1 | 3 - 4 | 1047 | 65 | ||
NM_057915.5 | NP_477263.1 | 3 - 4 | 1132 | 65 | ||
GenBank mRNA | AF151115.1 | - | 3 - 4 | 1110 | 65 | |
AY051877.1 | - | 3 - 4 | 1104 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
