Assay Details
Gene Symbol: Abi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104315.2 | NP_001097785.1 | 3 - 4 | 1213 | 67 | |
NM_001260161.1 | NP_001247090.1 | 4 - 5 | 1131 | 67 | ||
NM_001260162.1 | NP_001247091.1 | 4 - 5 | 79 | |||
NM_001275630.1 | NP_001262559.1 | 4 - 5 | 1216 | 67 | ||
NM_001275631.1 | NP_001262560.1 | 4 - 5 | 1134 | 67 | ||
NM_057915.5 | NP_477263.1 | 4 - 5 | 1219 | 67 | ||
GenBank mRNA | AF151115.1 | - | 4 - 5 | 1197 | 67 | |
AY051877.1 | - | 4 - 5 | 1191 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
