Assay Details
Gene Symbol: Dl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260264.2 | NP_001247193.1 | 4 - 5 | 1364 | 57 | |
NM_057916.4 | NP_477264.1 | 4 - 5 | 1364 | 57 | ||
NM_169853.3 | NP_732412.1 | 4 - 5 | 1364 | 57 | ||
GenBank mRNA | BT003267.1 | - | 4 - 5 | 1336 | 57 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
