Assay Details
| Gene Symbol: Doa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014678.3 | NP_001014678.1 | 9 - 10 | 2854 | 59 | |
| NM_001014679.2 | NP_001014679.1 | 7 - 8 | 2217 | 59 | ||
| NM_001014680.4 | NP_001014680.1 | 10 - 11 | 1570 | 59 | ||
| NM_001014681.4 | NP_001014681.2 | 9 - 10 | 5835 | 59 | ||
| NM_001014682.3 | NP_001014682.1 | 10 - 11 | 1446 | 59 | ||
| NM_001043298.2 | NP_001036763.2 | 8 - 9 | 4280 | 59 | ||
| NM_001043299.3 | NP_001036764.2 | 11 - 12 | 1653 | 59 | ||
| NM_001043300.3 | NP_001036765.1 | 10 - 11 | 1570 | 59 | ||
| NM_001144648.3 | NP_001138120.1 | 11 - 12 | 1513 | 59 | ||
| NM_001144649.2 | NP_001138121.1 | 11 - 12 | 1513 | 59 | ||
| NM_001144650.1 | NP_001138122.1 | 10 - 11 | 1446 | 59 | ||
| NM_001260424.1 | NP_001247353.1 | 8 - 9 | 2491 | 59 | ||
| NM_001260425.1 | NP_001247354.1 | 9 - 10 | 2482 | 59 | ||
| NM_001382085.1 | NP_001368951.1 | 11 - 12 | 1710 | 59 | ||
| NM_176581.4 | NP_788758.3 | 9 - 10 | 3244 | 59 | ||
| GenBank mRNA | AY061474.1 | - | 10 - 11 | 1564 | 59 | |
| AY069573.1 | - | 7 - 8 | 2217 | 59 | ||
| BT133441.1 | - | 9 - 10 | 2852 | 59 | ||
| EF489455.1 | - | 9 - 10 | 2852 | 59 | ||
More Information
Set Membership: |
|

Doa
Genomic Map