Assay Details
| Gene Symbol: Doa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014678.3 | NP_001014678.1 | 11 - 12 | 3158 | 63 | |
| NM_001014679.2 | NP_001014679.1 | 9 - 10 | 2539 | 63 | ||
| NM_001014680.4 | NP_001014680.1 | 12 - 13 | 1874 | 63 | ||
| NM_001014681.4 | NP_001014681.2 | 11 - 12 | 6157 | 63 | ||
| NM_001014682.3 | NP_001014682.1 | 12 - 13 | 1750 | 63 | ||
| NM_001043298.2 | NP_001036763.2 | 10 - 11 | 4602 | 63 | ||
| NM_001043299.3 | NP_001036764.2 | 13 - 14 | 1957 | 63 | ||
| NM_001043300.3 | NP_001036765.1 | 12 - 13 | 1874 | 63 | ||
| NM_001144648.3 | NP_001138120.1 | 13 - 14 | 1817 | 63 | ||
| NM_001144649.2 | NP_001138121.1 | 13 - 14 | 1817 | 63 | ||
| NM_001144650.1 | NP_001138122.1 | 12 - 13 | 1750 | 63 | ||
| NM_001260424.1 | NP_001247353.1 | 10 - 11 | 2813 | 63 | ||
| NM_001260425.1 | NP_001247354.1 | 11 - 12 | 2804 | 63 | ||
| NM_001382085.1 | NP_001368951.1 | 13 - 14 | 2014 | 63 | ||
| NM_176581.4 | NP_788758.3 | 11 - 12 | 3566 | 63 | ||
| GenBank mRNA | AY061474.1 | - | 12 - 13 | 1868 | 63 | |
| AY069573.1 | - | 9 - 10 | 2539 | 63 | ||
| BT133441.1 | - | 11 - 12 | 3156 | 63 | ||
| EF489455.1 | - | 11 - 12 | 3156 | 63 | ||
More Information
Set Membership: |
|

Doa
Genomic Map