Assay Details
| Gene Symbol: Doa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014680.4 | NP_001014680.1 | 2 - 3 | 588 | 67 | |
| NM_001014682.3 | NP_001014682.1 | 2 - 3 | 464 | 67 | ||
| NM_001043299.3 | NP_001036764.2 | 2 - 3 | 464 | 67 | ||
| NM_001043300.3 | NP_001036765.1 | 2 - 3 | 588 | 67 | ||
| NM_001144648.3 | NP_001138120.1 | 3 - 4 | 531 | 67 | ||
| NM_001144649.2 | NP_001138121.1 | 3 - 4 | 531 | 67 | ||
| NM_001144650.1 | NP_001138122.1 | 2 - 3 | 464 | 67 | ||
| NM_001382085.1 | NP_001368951.1 | 2 - 3 | 464 | 67 | ||
| GenBank mRNA | AY061474.1 | - | 2 - 3 | 582 | 67 | |
More Information
Set Membership: |
|

Doa
Genomic Map