Assay Details
Gene Symbol: Mkk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170080.2 | NP_001163551.1 | 1 - 2 | 315 | 98 | |
NM_001275446.1 | NP_001262375.1 | 1 - 2 | 315 | 98 | ||
NM_058005.5 | NP_477353.1 | 1 - 2 | 315 | 98 | ||
GenBank mRNA | AY070600.1 | - | 1 - 2 | 313 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
