Assay Details
| Gene Symbol: Gld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001363924.1 | NP_001350861.1 | 3 - 4 | 637 | 77 | |
| NM_058155.5 | NP_477503.1 | 3 - 4 | 637 | 77 | ||
| GenBank mRNA | BT150358.1 | - | 3 - 4 | 637 | 77 | |
More Information
Set Membership: |
|

Gld
Genomic Map