Assay Details
| Gene Symbol: Dlip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079500.4 | NP_524224.2 | 1 - 2 | 479 | 66 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlip2| Gene Symbol: Dlip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079500.4 | NP_524224.2 | 1 - 2 | 479 | 66 | |
Set Membership: | 
		   
					
						
							 |