Assay Details
| Gene Symbol: Ubc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259987.3 | NP_001246916.1 | 1 - 2 | 227 | 65 | |
| NM_079506.4 | NP_524230.2 | 1 - 2 | 227 | 65 | ||
| GenBank mRNA | BT003481.1 | - | 1 - 2 | 229 | 65 | |
| KX531446.1 | - | 1 - 2 | 41 | 65 | ||
More Information
Set Membership: |
|

Ubc6
Genomic Map