Assay Details
Gene Symbol: Vha26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300253.1 | NP_001287182.1 | 2 - 3 | 495 | 90 | |
NM_079513.3 | NP_524237.1 | 3 - 4 | 397 | 90 | ||
NM_169073.2 | NP_730957.1 | 2 - 3 | 495 | 90 | ||
GenBank mRNA | AF145618.1 | - | 2 - 3 | 440 | 90 | |
U38198.1 | - | 2 - 3 | 329 | 90 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
