Assay Details
Gene Symbol: Obp83a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300260.1 | NP_001287189.1 | 3 - 4 | 471 | 57 | |
NM_001300261.1 | NP_001287190.1 | 3 - 4 | 409 | 57 | ||
NM_079517.3 | NP_524241.1 | 3 - 4 | 425 | 57 | ||
GenBank mRNA | BT029030.1 | - | 3 - 4 | 409 | 57 | |
U02542.1 | - | 3 - 4 | 419 | 57 | ||
U05982.1 | - | 3 - 4 | 424 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
