Assay Details
| Gene Symbol: NPFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260016.2 | NP_001246945.1 | 3 - 4 | 891 | 65 | |
| NM_001260017.2 | NP_001246946.1 | 3 - 4 | 891 | 65 | ||
| NM_001260018.2 | NP_001246947.1 | 3 - 4 | 891 | 65 | ||
| NM_079521.4 | NP_524245.3 | 3 - 4 | 891 | 65 | ||
| GenBank mRNA | BT044552.1 | - | 3 - 4 | 806 | 65 | |
| DQ666851.1 | - | 3 - 4 | 744 | 65 | ||
More Information
Set Membership: |
|

NPFR
Genomic Map