Assay Details
| Gene Symbol: NPFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260016.2 | NP_001246945.1 | 4 - 5 | 1066 | 72 | |
| NM_001260017.2 | NP_001246946.1 | 4 - 5 | 1066 | 72 | ||
| NM_001260018.2 | NP_001246947.1 | 4 - 5 | 1066 | 72 | ||
| NM_079521.4 | NP_524245.3 | 4 - 5 | 1066 | 72 | ||
| GenBank mRNA | BT044552.1 | - | 4 - 5 | 981 | 72 | |
| DQ666851.1 | - | 4 - 5 | 919 | 72 | ||
More Information
Set Membership: |
|

NPFR
Genomic Map