Assay Details
| Gene Symbol: Scr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001369958.1 | NP_001356915.1 | 2 - 3 | 1566 | 57 | |
| NM_001369959.1 | NP_001356897.1 | 2 - 3 | 1566 | 57 | ||
| NM_001382008.1 | NP_001368995.1 | 2 - 3 | 1566 | 57 | ||
| NM_079524.4 | NP_524248.2 | 2 - 3 | 1566 | 57 | ||
| NM_206443.3 | NP_996165.1 | 2 - 3 | 1413 | 57 | ||
More Information
Set Membership: |
|

Scr
Genomic Map