Assay Details
Gene Symbol: lap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260029.2 | NP_001246958.1 | 8 - 9 | 1368 | 73 | |
NM_001260030.2 | NP_001246959.1 | 8 - 9 | 1368 | 73 | ||
NM_001260031.2 | NP_001246960.1 | 8 - 9 | 1368 | 73 | ||
NM_079528.3 | NP_524252.2 | 8 - 9 | 1368 | 73 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
