Assay Details
Gene Symbol: lap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144547.3 | NP_001138019.2 | 9 - 10 | 1528 | 104 | |
NM_001170067.2 | NP_001163538.1 | 9 - 10 | 1528 | 104 | ||
NM_001260029.2 | NP_001246958.1 | 9 - 10 | 1480 | 104 | ||
NM_001260030.2 | NP_001246959.1 | 9 - 10 | 1480 | 104 | ||
NM_001260031.2 | NP_001246960.1 | 9 - 10 | 1480 | 104 | ||
NM_001275409.1 | NP_001262338.1 | 8 - 9 | 1495 | 104 | ||
NM_001275410.1 | NP_001262339.1 | 9 - 10 | 1528 | 104 | ||
NM_001275411.1 | NP_001262340.1 | 8 - 9 | 1495 | 104 | ||
NM_079528.3 | NP_524252.2 | 9 - 10 | 1480 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
