Assay Details
| Gene Symbol: alpha-Est7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079537.4 | NP_524261.1 | 5 - 6 | 1347 | 70 | |
| GenBank mRNA | AY051473.1 | - | 5 - 6 | 1327 | 70 | |
More Information
Set Membership: |
|

alpha-Est7
Genomic Map