Assay Details
Gene Symbol: dsx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300291.1 | NP_001287220.1 | 3 - 4 | 2458 | 58 | |
NM_079548.5 | NP_524272.4 | 3 - 4 | 2069 | 58 | ||
NM_169203.2 | NP_731198.1 | 3 - 4 | 2458 | 58 | ||
GenBank mRNA | BT029258.1 | - | 3 - 4 | 2069 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
