Assay Details
| Gene Symbol: mRpL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079551.4 | NP_524275.1 | 1 - 2 | 268 | 75 | |
| GenBank mRNA | AY051800.1 | - | 1 - 2 | 249 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mRpL1| Gene Symbol: mRpL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079551.4 | NP_524275.1 | 1 - 2 | 268 | 75 | |
| GenBank mRNA | AY051800.1 | - | 1 - 2 | 249 | 75 | |
Set Membership: |
|