Assay Details
| Gene Symbol: RhoL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260074.1 | NP_001247003.1 | 3 - 4 | 731 | 66 | |
| NM_079568.3 | NP_524292.1 | 3 - 4 | 661 | 66 | ||
| GenBank mRNA | AY069811.1 | - | 3 - 4 | 645 | 66 | |
| BT120283.1 | - | 3 - 4 | 731 | 66 | ||
| U50314.1 | - | 3 - 4 | 661 | 66 | ||
More Information
Set Membership: |
|

RhoL
Genomic Map