Assay Details
| Gene Symbol: hyd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300333.1 | NP_001287262.1 | 11 - 12 | 5852 | 82 | |
| NM_079572.4 | NP_524296.2 | 11 - 12 | 5846 | 82 | ||
| GenBank mRNA | BT150484.1 | - | 11 - 12 | 5830 | 82 | |
More Information
Set Membership: |
|

hyd
Genomic Map