Assay Details
| Gene Symbol: hyd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300333.1 | NP_001287262.1 | 12 - 13 | 6593 | 130 | |
| NM_079572.4 | NP_524296.2 | 12 - 13 | 6587 | 130 | ||
| GenBank mRNA | BT150484.1 | - | 12 - 13 | 6571 | 130 | |
More Information
Set Membership: |
|

hyd
Genomic Map