Assay Details
| Gene Symbol: hyd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300333.1 | NP_001287262.1 | 17 - 18 | 8310 | 101 | |
| NM_079572.4 | NP_524296.2 | 17 - 18 | 8304 | 101 | ||
| GenBank mRNA | BT150484.1 | - | 17 - 18 | 8288 | 101 | |
More Information
Set Membership: |
|

hyd
Genomic Map