Assay Details
Gene Symbol: Timp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169336.4 | NP_731461.1 | 4 - 5 | 762 | 61 | |
GenBank mRNA | AY069211.1 | - | 4 - 5 | 743 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
