Assay Details
Gene Symbol: mRpL37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079582.5 | NP_524306.3 | 3 - 4 | 894 | 67 | |
GenBank mRNA | BT030992.1 | - | 3 - 4 | 874 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
