Assay Details
Gene Symbol: RpL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079592.3 | NP_524316.1 | 2 - 3 | 230 | 52 | |
NM_169377.2 | NP_731547.1 | 2 - 3 | 230 | 52 | ||
NM_169378.2 | NP_731548.2 | 2 - 3 | 230 | 52 | ||
NM_169380.2 | NP_731550.2 | 2 - 3 | 230 | 52 | ||
GenBank mRNA | AF016835.1 | - | 2 - 3 | 227 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
