Assay Details
Gene Symbol: Ho | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079597.3 | NP_524321.1 | 2 - 3 | 787 | 67 | |
GenBank mRNA | AY061087.1 | - | 2 - 3 | 761 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
