Assay Details
Gene Symbol: Prosalpha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079604.3 | NP_524328.1 | 2 - 3 | 243 | 83 | |
GenBank mRNA | AY069280.1 | - | 2 - 3 | 181 | 83 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
