Assay Details
Gene Symbol: Prosalpha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079604.3 | NP_524328.1 | 4 - 5 | 652 | 60 | |
GenBank mRNA | AY069280.1 | - | 4 - 5 | 590 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
