Assay Details
| Gene Symbol: Paip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275586.1 | NP_001262515.1 | 3 - 4 | 483 | 82 | |
| NM_079610.3 | NP_524334.1 | 2 - 3 | 497 | 82 | ||
| GenBank mRNA | AY061203.1 | - | 2 - 3 | 449 | 82 | |
| AY333418.1 | - | 2 - 3 | 497 | 82 | ||
| BT124998.1 | - | 2 - 3 | 428 | 82 | ||
| KX531722.1 | - | 1 - 2 | 150 | 82 | ||
More Information
Set Membership: |
|

Paip2
Genomic Map