Assay Details
Gene Symbol: CtBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014617.2 | NP_001014617.1 | 2 - 3 | 576 | 78 | |
NM_001275591.2 | NP_001262520.1 | 2 - 3 | 576 | 78 | ||
NM_001275592.1 | NP_001262521.1 | 2 - 3 | 563 | 78 | ||
NM_001275593.1 | NP_001262522.1 | 2 - 3 | 576 | 78 | ||
NM_001275594.1 | NP_001262523.1 | 2 - 3 | 576 | 78 | ||
NM_001275595.1 | NP_001262524.1 | 2 - 3 | 576 | 78 | ||
NM_079612.3 | NP_524336.2 | 2 - 3 | 576 | 78 | ||
NM_169490.2 | NP_731762.1 | 2 - 3 | 716 | 78 | ||
NM_169491.2 | NP_731763.1 | 2 - 3 | 1000 | 78 | ||
NM_169492.2 | NP_731764.1 | 2 - 3 | 563 | 78 | ||
GenBank mRNA | AB011840.1 | - | 1 - 2 | 198 | 78 | |
AY069170.1 | - | 2 - 3 | 716 | 78 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
