Assay Details
| Gene Symbol: ry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079613.3 | NP_524337.1 | 2 - 3 | 2882 | 82 | |
| GenBank mRNA | BT015293.1 | - | 2 - 3 | 2860 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ry| Gene Symbol: ry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079613.3 | NP_524337.1 | 2 - 3 | 2882 | 82 | |
| GenBank mRNA | BT015293.1 | - | 2 - 3 | 2860 | 82 | |
Set Membership: |
|