Assay Details
| Gene Symbol: ry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079613.3 | NP_524337.1 | 3 - 4 | 4197 | 74 | |
| GenBank mRNA | BT015293.1 | - | 3 - 4 | 4175 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ry| Gene Symbol: ry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079613.3 | NP_524337.1 | 3 - 4 | 4197 | 74 | |
| GenBank mRNA | BT015293.1 | - | 3 - 4 | 4175 | 74 | |
Set Membership: |
|