Assay Details
| Gene Symbol: sim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275598.2 | NP_001262527.1 | 2 - 3 | 406 | 69 | |
| NM_001275599.1 | NP_001262528.1 | 2 - 3 | 686 | 69 | ||
| NM_079616.4 | NP_524340.2 | 2 - 3 | 349 | 69 | ||
| NM_169495.4 | NP_731771.3 | 2 - 3 | 686 | 69 | ||
| GenBank mRNA | AY129457.1 | - | 2 - 3 | 346 | 69 | |
More Information
Set Membership: |
|

sim
Genomic Map