Assay Details
Gene Symbol: timeout | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079617.3 | NP_524341.3 | 3 - 4 | 792 | 92 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
